Läs mer
(7Alpha,8Beta,9Alpha,10Alpha)-7,8,9,10-Tetrahydrobenzo[a]pyrene-7,8,9,10-tetrol (>90%), TRC
Produktkod. 30133467
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30133467 5 mg -
30007195 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133467 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3100005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61490-69-5
Molekylformel C20 H16 O4
LEDER O[C@@H]1[C@@H](O)[C@H](O)c2cc3ccc4cccc5ccc(c2[C@@H]1O)c3c45
Specifikationer
Kemiskt namn eller material Benzo[a]pyrenetetrol II 2
Kvantitet 5 mg
Synonym Benzo[a]pyrene-7,8,9,10-tetrol, 7,8,9,10-tetrahydro-, (7R,8S,9S,10S)-rel-, Benzo[a]pyrene-7,8,9,10-tetrol, 7,8,9,10-tetrahydro-, (7α,8β,9α,10α)-, rel-(7R,8S,9S,10S)-7,8,9,10-Tetrahydrobenzo[a]pyrene-7,8,9,10-tetrol, (±)-Benzo[a]pyrene-r-7,t-8,c-9,c-10-tetrahydrotetrol, 7,9,10/8-Tetrahydroxytetrahydrobenzo[a]pyrene, 7β,8α,9β,10β-Tetrahydroxy-7,8,9,10-tetrahydrobenzo[a]pyrene, Benzo[a]pyrenetetrol II 2, Tetrol II 2
InChI formel InChI=1S/C20H16O4/c21-17-13-8-11-5-4-9-2-1-3-10-6-7-12(15(11)14(9)10)16(13)18(22)20(24)19(17)23/h1-8,17-24H/t17-,18+,19+,20+/m1/s1
IUPAC-namn (7R,8S,9S,10S)-7,8,9,10-tetrahydrobenzo[a]pyrene-7,8,9,10-tetrol
Molekylvikt (g/mol) 320.34
Formel vikt 320.1049