Läs mer
7Beta-Spironolactone, TRC
Produktkod. 30418646
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418646 10 mg
30411736 25 mg
30409511 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30418646 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68301010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 33784-05-3
Molekylformel C24 H32 O4 S
Molekylvikt (g/mol) 416.57
Synonym Pregn-4-ene-21-carboxylic acid, 7-(acetylthio)-17-hydroxy-3-oxo-, γ-lactone, (7β,17α)-, 17α-Pregn-4-ene-21-carboxylic acid, 17-hydroxy-7β-mercapto-3-oxo-, γ-lactone, acetate (8CI), 3'-(3-Oxo-7β-acetylthio-17β-hydroxyandrost-4-en-17α-yl)-propionic acid lactone, 7β-(Acetylthio)-17α-hydroxy-3-oxopregn-4-ene-21-carboxylic acid γ-lactone, SC 11940
IUPAC-namn S-[(7S,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
LEDER CC(=O)S[C@H]1CC2=CC(=O)CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]45CCC(=O)O5)[C@H]13
Specifikationer
Kemiskt namn eller material 7-beta-Spironolactone
Kvantitet 10 mg
InChI formel InChI=1S/C24H32O4S/c1-14(25)29-19-13-15-12-16(26)4-8-22(15,2)17-5-9-23(3)18(21(17)19)6-10-24(23)11-7-20(27)28-24/h12,17-19,21H,4-11,13H2,1-3H3/t17-,18-,19-,21+,22-,23-,24+/m0/s1
Formel vikt 416.2021