Läs mer
(7Z)-7-Hexadecenoic Acid (Solution in Ethanol), TRC
Produktkod. 30405551
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30405551 10 mg
30404186 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30405551 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29354010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (7Z)-7-Hexadecenoic Acid
CAS 2416-19-5
Molekylformel C16 H30 O2
Kvantitet 10 mg
Synonym (Z)-7-Hexadecenoic acid, 7Z-Hexadecenoic acid, Hypogeic acid, cis-7-Hexadecenoic acid
InChI formel InChI=1S/C16H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h9-10H,2-8,11-15H2,1H3,(H,17,18)/b10-9-
LEDER CCCCCCCC\C=C/CCCCCC(=O)O
IUPAC-namn (Z)-hexadec-7-enoic acid
Molekylvikt (g/mol) 254.41
Formel vikt 254.2246