missing translation for 'onlineSavingsMsg'
Läs mer

8-(2-Pyrimidinyl)-8-aza-5-azoniaspiro[4.5]decane Bromide, TRC

Produktkod. 30427952
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427952 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30427952 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP997235500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 8-(Pyrimidin-2-yl)-8-aza-5-azoniaspiro[4.5]decane Bromide
CAS 81461-73-6
Molekylformel C12 H19 N4 . Br
Kvantitet 500 mg
Synonym 8-Aza-5-azoniaspiro[4.5]decane','8-(2-pyrimidinyl)-','bromide (1:1), 8-Aza-5-azoniaspiro[4.5]decane','8-(2-pyrimidinyl)-','bromide (9CI), 8-(2-Pyrimidinyl)-8-aza-5-azoniaspiro[4.5]decane bromide
InChI formel InChI=1S/C12H19N4.BrH/c1-2-9-16(8-1)10-6-15(7-11-16)12-13-4-3-5-14-12;/h3-5H,1-2,6-11H2;1H/q+1;/p-1
LEDER [Br-].C1CC[N+]2(C1)CCN(CC2)c3ncccn3
IUPAC-namn 8-pyrimidin-2-yl-8-aza-5-azoniaspiro[4.5]decane;bromide
Molekylvikt (g/mol) 299.2101
Formel vikt 298.0793
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.