Learn More
8-Anilino-1-naphthalenesulfonic Acid Hemimagnesium Salt, TRC
Product Code. 30337355
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
30337355 10 g 10g
1 options
Product Code. 30337355 Supplier Toronto Research Chemicals Supplier No. TRCA66379810G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 8-Anilino-1-naphthalenesulfonic Acid Hemimagnesium Salt
CAS 18108-68-4
Molecular Formula 2[C16 H12 N O3 S] . Mg
Quantity 10 g
Synonym 1-Anilino-8-naphthalenesulfonate Magnesium Salt, 1-Anilino-8-naphthalenesulfonic Acid Magnesium Salt, 8-Anilino-1-naphthalenesulfonate Magnesium Salt, 8-Anilinonaphthalene-1-sulfonic acid Magnesium Salt, ANS hemi-Mg Salt, Magnesium 1-Anilino-8-naphthalenesulfonate, Magnesium 8-Anilino-1-naphthalenesulfonate
InChI Formula InChI=1S/2C16H13NO3S.Mg/c2*18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13;/h2*1-11,17H,(H,18,19,20);/q;;+2/p-2
SMILES [Mg+2].[O-]S(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12.[O-]S(=O)(=O)c4cccc5cccc(Nc6ccccc6)c45
IUPAC Name magnesium;8-anilinonaphthalene-1-sulfonate
Molecular Weight (g/mol) 620.978
Formula Weight 620.093