Läs mer
8-Bromo-3,9-dihydro-3-methyl-1H-purine-2,6-dione, TRC
Produktkod. 30011058
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011058 250 mg -
30008711 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011058 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB683485250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93703-24-3
Molekylformel C6H5BrN4O2
Molekylvikt (g/mol) 245.03
Synonym 3-Methyl-8-bromoxanthine, 8-Bromo-3-methyl-3,7-dihydropurine-2,6-dione, 8-Bromo-3-methylxanthine, Linagliptin 8-Bromo Impurity
IUPAC-namn 8-bromo-3-methyl-7H-purine-2,6-dione
LEDER CN1C(=O)NC(=O)c2[nH]c(Br)nc12
Specifikationer
Kemiskt namn eller material 8-Bromo-3,9-dihydro-3-methyl-1H-purine-2,6-dione
Kvantitet 250 mg
InChI formel InChI=1S/C6H5BrN4O2/c1-11-3-2(8-5(7)9-3)4(12)10-6(11)13/h1H3,(H,8,9)(H,10,12,13)
Formel vikt 243.9596