Läs mer
8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione, TRC
Produktkod. 30009826
Change view
Kvantitet:
1 g
2.5 g
10 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009826 1 g -
30004717 2.5 g 2.5g
30009194 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009826 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6805101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 853029-57-9
Molekylformel C20 H17 Br N6 O2
Molekylvikt (g/mol) 453.29
Synonym 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione, 1H-Purine-2,6-dione, 8-bromo-7-(2-butynyl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]- (9CI), 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione, 1-[(4-Methylquinazolin-2-yl)methyl]-3-methyl-7-(2-butyn-1-yl)-8-bromoxanthine
IUPAC-namn 8-bromo-7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]purine-2,6-dione
LEDER CC#CCn1c(Br)nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c12
Specifikationer
Kemiskt namn eller material 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C20H17BrN6O2/c1-4-5-10-26-16-17(24-19(26)21)25(3)20(29)27(18(16)28)11-15-22-12(2)13-8-6-7-9-14(13)23-15/h6-9H,10-11H2,1-3H3
Formel vikt 452.0596
Procent renhet >95%