Learn More
IBA Solutions For Life Science 8-Bromo-GTP
A modified nucleoside-triphosphate (modified 2´-ribonucleoside-5´-triphosphates) that helps in studying RNA-protein interactions.
Brand:  IBA Solutions For Life Science 5-0613-514
Additional Details : Weight : 0.00050kg
Product Code. 15653648
  • 2053.00 SEK / 1µmol
Estimated Shipment
03-10-2019
Add to basket
Description
Description
IBA has a nucleoside-triphosphate synthesis facility that offers an extensive range of triphosphates for many applications.
  • All products are double HPLC purified, desalted and well characterized by HPLC and 31P-NMR
  • They are also available as monophosphates and nucleosides
Specifications
Specifications
Yes
Helps in studying RNA-protein interactions
1.0 μmol
Yes
Nucleoside-triphosphate