missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenosine-3',5'-cyclic monophosphate, Thermo Scientific Chemicals

Catalog Number p-7042938
Click to view available options
Quantity:
50 mg
100 mg
250 mg
This item is not returnable. View return policy
23583-48-4
C10H11BrN5O6P
408.11
MFCD00075580
DVKQVRZMKBDMDH-UUOKFMHZSA-N
bcamp, 8-bromo-cyclic amp, 8-br-camp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromo-cyclic 3',5'-amp, 8-bromo-camp, 8-bromo cyclic adenosine monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, cyclic 8-bromoadenosine 3',5'-monophosphate
32014
CHEBI:64211
(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
NC1=C2N=C(Br)N([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1
This item is not returnable. View return policy
23583-48-4
C10H11BrN5O6P
408.11
MFCD00075580
DVKQVRZMKBDMDH-UUOKFMHZSA-N
bcamp, 8-bromo-cyclic amp, 8-br-camp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromo-cyclic 3',5'-amp, 8-bromo-camp, 8-bromo cyclic adenosine monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, cyclic 8-bromoadenosine 3',5'-monophosphate
32014
CHEBI:64211
(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
NC1=C2N=C(Br)N([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 254°C
Color Brown
Quantity 50 mg
Sensitivity Hygroscopic
Formula Weight 408.11
Physical Form Powder
Chemical Name or Material 8-Bromoadenosine-3',5'-cyclic monophosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'