Läs mer
8-chloro-2,3-dihydro-1,4-benzodioxine-5-carboxylic Acid, TRC
Produktkod. 30450693
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30450693 2.5 mg 2.5mg
30450082 5 mg -
30449247 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450693 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7766182.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66411-24-3
Molekylformel C9H7O4Cl
Molekylvikt (g/mol) 214.6
IUPAC-namn 5-chloro-2,3-dihydro-1,4-benzodioxine-8-carboxylic acid
LEDER OC(=O)c1ccc(Cl)c2OCCOc12
Specifikationer
Kemiskt namn eller material 8-chloro-2,3-dihydro-1,4-benzodioxine-5-carboxylic Acid
Kvantitet 2.5 mg
InChI formel InChI=1S/C9H7ClO4/c10-6-2-1-5(9(11)12)7-8(6)14-4-3-13-7/h1-2H,3-4H2,(H,11,12)
Formel vikt 214.0033