missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline, 97%, Thermo Scientific™

Catalog Number p-7024717
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
18706-22-4
C10H5ClF3NO
247.601
MFCD00276589
SBVZSZNYPRWWNN-UHFFFAOYSA-N
8-chloro-4-hydroxy-2-trifluoromethyl quinoline, 8-chloro-2-trifluoromethyl quinolin-4-ol, 8-chloro-2-trifluoromethyl-1h-quinolin-4-one, 8-chloro-2-trifluoromethyl-4-quinolinol, 4 1h-quinolinone, 8-chloro-2-trifluoromethyl, maybridge1_004966, pubchem5940, acmc-1bvi2, 8-chloro-2-trifluoromethyl-quinolin-4-ol, 8-chloro-4-hydroxy-2-trifluoromethylquinoline
2736611
8-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)Cl)NC(=CC2=O)C(F)(F)F
This item is not returnable. View return policy
18706-22-4
C10H5ClF3NO
247.601
MFCD00276589
SBVZSZNYPRWWNN-UHFFFAOYSA-N
8-chloro-4-hydroxy-2-trifluoromethyl quinoline, 8-chloro-2-trifluoromethyl quinolin-4-ol, 8-chloro-2-trifluoromethyl-1h-quinolin-4-one, 8-chloro-2-trifluoromethyl-4-quinolinol, 4 1h-quinolinone, 8-chloro-2-trifluoromethyl, maybridge1_004966, pubchem5940, acmc-1bvi2, 8-chloro-2-trifluoromethyl-quinolin-4-ol, 8-chloro-4-hydroxy-2-trifluoromethylquinoline
2736611
8-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C(=C1)Cl)NC(=CC2=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 138°C to 140°C
Quantity 1 g
Formula Weight 247.61
Percent Purity 97%
Chemical Name or Material 8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'