Läs mer
8-CPT-cAMP sodium salt, TRC
Produktkod. 30352856
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30352856 5 mg
1 options
Produktkod. 30352856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC7814805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 8-CPT-cAMP sodium salt
CAS 93882-12-3
Molekylformel C16H15ClN5O6PS • Na
Kvantitet 5 mg
Synonym Adenosine, 8-[(4-chlorophenyl)thio]-, cyclic 3',5'-(hydrogen phosphate), sodium salt (1:1) (ACI), 4H-Furo[3,2-d]-1,3,2-dioxaphosphorin, adenosine deriv. (ZCI), Adenosine, 8-[(4-chlorophenyl)thio]-, cyclic 3',5'-(hydrogen phosphate), monosodium salt (9CI), 8-(p-Chlorophenylthio)-cAMP sodium salt, 8-CPT-cAMP sodium salt, 8-[(4-Chlorophenyl)thio]adenosine cyclic 3',5'-(hydrogen phosphate) monosodium salt
InChI formel InChI=1S/C16H15ClN5O6PS.Na/c17-7-1-3-8(4-2-7)30-16-21-10-13(18)19-6-20-14(10)22(16)15-11(23)12-9(27-15)5-26-29(24,25)28-12;/h1-4,6,9,11-12,15,23H,5H2,(H,24,25)(H2,18,19,20);/q;+1/p-1/t9-,11-,12-,15-;/m1./s1
LEDER ClC(C=C1)=CC=C1SC2=NC3=C(N)N=CN=C3N2C4[C@H](O)[C@@](OP(OC5)(O)=O)([H])[C@]5([H])O4.[Na]
IUPAC-namn sodium;(4aR,6R,7R,7aS)-6-[6-amino-8-(4-chlorophenyl)sulfanylpurin-9-yl]-2-oxido-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
Molekylvikt (g/mol) 471.812299
Formel vikt 492.9989