Läs mer
8-Desethyl Etodolac, TRC
Produktkod. 30363858
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363858 250 mg
30360863 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363858 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289315250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41339-67-7
Molekylformel C15 H17 N O3
Molekylvikt (g/mol) 259.3
Synonym 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid, 8-Desethyl Etodolac, Pyrano[3,4-b]indole-1-acetic acid, 1-ethyl-1,3,4,9-tetrahydro-, 1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid
IUPAC-namn 2-(1-ethyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid
LEDER CCC1(CC(=O)O)OCCc2c1[nH]c3ccccc23
Specifikationer
Kemiskt namn eller material 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac)
Kvantitet 250 mg
InChI formel InChI=1S/C15H17NO3/c1-2-15(9-13(17)18)14-11(7-8-19-15)10-5-3-4-6-12(10)16-14/h3-6,16H,2,7-9H2,1H3,(H,17,18)
Formel vikt 259.1208