Läs mer
8-Ethyl Irinotecan, TRC
Produktkod. 30374956
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374956 25 mg
30384548 1 mg
30375801 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30374956 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92110025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 947687-02-7
Molekylformel C35 H42 N4 O6
Molekylvikt (g/mol) 614.73
Synonym (4S)-4,8,11-Triethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-Bipiperidine]-1'-carboxylic Acid Ester, (4S)-4,8,11-Triethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-Bipiperidine]-1'-carboxylate, (4S)-4,8,11-triethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinolin-9-yl 1,4′-bipiperidine-1′-carboxylate, 8-Ethyl Irinotecan, Irinotecan Hydrochloride Trihydrate Imp. C (EP)
LEDER CCc1cc2nc3C4=CC5=C(COC(=O)[C@]5(O)CC)C(=O)N4Cc3c(CC)c2cc1OC(=O)N6CCC(CC6)N7CCCCC7
Specifikationer
Kemiskt namn eller material 8-ethyl Irinotecan
Kvantitet 25 mg
InChI formel InChI=1S/C35H42N4O6/c1-4-21-16-28-24(17-30(21)45-34(42)38-14-10-22(11-15-38)37-12-8-7-9-13-37)23(5-2)25-19-39-29(31(25)36-28)18-27-26(32(39)40)20-44-33(41)35(27,43)6-3/h16-18,22,43H,4-15,19-20H2,1-3H3/t35-/m0/s1
Formel vikt 614.3104