Läs mer
8-Hydroxy-5-quinolinesulfonic acid, TRC
Produktkod. 30406807
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30406807 25 g
1 options
Produktkod. 30406807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95317525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 8-Hydroxy-5-quinolinesulfonic acid
CAS 84-88-8
Molekylformel C9 H7 N O4 S
Kvantitet 25 g
Synonym 8-Hydroxy-5-quinolinesulfonic acid, 5-Sulfo-8-hydroxyquinoline, 5-Sulfo-8-quinolinol, 5-Sulfooxine, 8-Hydroxy-5-quinolinesulfonic acid, 8-Hydroxy-5-sulfoquinoline, 8-Hydroxylquinoline-5-sulfonic acid, 8-Hydroxyquinoline-5-sulfonic acid, 8-Oxyquinoline-5-sulfonic acid, 8-Quinolinol-5-sulfonic acid, HQSA, NSC 105877, NSC 13139, NSC 220049, Oxine-5-sulfonic acid, Sulfoxine
InChI formel InChI=1S/C9H7NO4S/c11-7-3-4-8(15(12,13)14)6-2-1-5-10-9(6)7/h1-5,11H,(H,12,13,14)
LEDER Oc1ccc(c2cccnc12)S(=O)(=O)O
IUPAC-namn 8-hydroxyquinoline-5-sulfonic acid
Molekylvikt (g/mol) 225.22
Formel vikt 225.0096