Läs mer
8-Hydroxy Adenine, TRC
Produktkod. 30403255
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30403255 250 mg
30406282 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30403255 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH750060250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21149-26-8
Molekylformel C5H5N5O
Molekylvikt (g/mol) 151.13
Synonym 6-Amino-7,9-dihydro-8H-purin-8-one, 6-Amino-1,7-dihydro-8H-purin-8-one, 6-Amino-purin-8-ol, 6-Amino-8-hydroxypurine, 7,8-Dihydro-8-oxoadenine, 8-Oxo-7,8-dihydroadenine, 8-Oxoadenine, NSC 24123, Oksaden, Oxaden, Oxyadenine
IUPAC-namn 6-amino-9H-purin-8-ol
LEDER Nc1ncnc2[nH]c(O)nc12
Specifikationer
Kemiskt namn eller material 8-Hydroxy Adenine
Kvantitet 250 mg
InChI formel InChI=1S/C5H5N5O/c6-3-2-4(8-1-7-3)10-5(11)9-2/h1H,(H4,6,7,8,9,10,11)
Formel vikt 151.0494