Läs mer
8-Hydroxy Guanosine, TRC
Produktkod. 17954875
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17954875 10 mg
17225103 5 mg
17933965 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17954875 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94277010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3868-31-3
Molekylformel C10 H13 N5 O6
Molekylvikt (g/mol) 299.24
Synonym Guanosine, 7,8-dihydro-8-oxo-, Purine-6,8(1H,9H)-dione, 2-amino-9-β-D-ribofuranosyl- (7CI,8CI), 7,8-Dihydro-8-oxoguanosine, 8-Hydroxyguanosine, 8-Oxo-7,8-dihydroguanosine, 8-Oxoguanosine, Chembridge 5144558, NQZ-013, NSC 90393
IUPAC-namn 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-8-hydroxy-1H-purin-6-one
LEDER NC1=Nc2c(nc(O)n2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1
Specifikationer
Kemiskt namn eller material 8-Hydroxyguanosine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C10H13N5O6/c11-9-13-6-3(7(19)14-9)12-10(20)15(6)8-5(18)4(17)2(1-16)21-8/h2,4-5,8,16-18H,1H2,(H,12,20)(H3,11,13,14,19)/t2-,4-,5-,8-/m1/s1
Formel vikt 299.0866
Procent renhet >95%