Läs mer
8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt, Mikromol™
Produktkod. 17923586
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17923586 25 mg
17950696 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17923586 Leverantör Mikromol Leverantörsnummer MM0599.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
CAS 64487-94-1
Analyt- eller komponentnamn 8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
Frakt skick Room Temperature
Molekylformel C24 H27 N5 O3
Certifikat och efterlevnad ISO 17025
Synonym 8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
InChI formel InChI=1S/C24H27N5O3/c1-27-21-19(22(30)28(2)24(27)31)25-23(26-21)29(3,4)15-16-32-20(17-11-7-5-8-12-17)18-13-9-6-10-14-18/h5-14,20H,15-16H2,1-4H3
LEDER CN1C(=O)N(C)c2nc([n-]c2C1=O)[N+](C)(C)CCOC(c3ccccc3)c4ccccc4
IUPAC-namn 2-benzhydryloxyethyl-(1,3-dimethyl-2,6-dioxopurin-7-id-8-yl)-dimethylazanium
Molekylvikt (g/mol) 433.5
Kvantitet 25 mg
Formel vikt 433.2114 g/mol