Läs mer
8-Nitroguanine, TRC
Produktkod. 30464878
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464878 10 mg
30465966 5 mg
30467080 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49601010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 168701-80-2
Molekylformel C5 H4 N6 O3
Molekylvikt (g/mol) 196.12
IUPAC-namn 2-amino-8-nitro-1,7-dihydropurin-6-one
LEDER NC1=Nc2nc([nH]c2C(=O)N1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 8-Nitroguanine
Kvantitet 10 mg
InChI formel InChI=1S/C5H4N6O3/c6-4-8-2-1(3(12)10-4)7-5(9-2)11(13)14/h(H4,6,7,8,9,10,12)
Formel vikt 196.0345