Läs mer
8-OH-DPAT, MedChemExpress
Produktkod. 16487267
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16487267 5 mg
16450857 50 mg
16445406 100 mg
16433126 25 mg
16424558 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16487267 Leverantör Medchem Express Leverantörsnummer HY112061
MedChemExpress 8-OH-DPAT is a potent and selective 5-HT agonist, with a pIC50 of 8.19 for 5-HT1A and a Ki of 466 nM for 5-HT7; 8-OH-DPAT weakly binds to 5-HT1B (pIC50, 5.42), 5-HT (pIC50 <5).
8-OH-DPAT is a potent and selective 5-HT agonist, with a pIC50 of 8.19 for 5-HT1A and a Ki of 466 nM for 5-HT7
Specifikationer
Kemiskt namn eller material 8-OH-DPAT
CAS 78950-78-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C16H25NO
Kvantitet 5 mg
Synonym 8-Hydroxy-DPAT
Löslighetsinformation DMSO : 100 mg/mL (404.24 mM; Need ultrasonic)
LEDER OC1=C2CC(N(CCC)CCC)CCC2=CC=C1
Molekylvikt (g/mol) 247.38
Formel vikt 247.38
Procent renhet 98.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Neuroscience-Neuromodulation