Läs mer
8-Oxoadenosine, TRC
Produktkod. 30422484
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422484 10 mg
30421343 100 mg
30428221 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30422484 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO84726310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29851-57-8
Molekylformel C10H13N5O5
Molekylvikt (g/mol) 283.24
Synonym NSC 102240, NSC 90396, 7,8-Dihydro-8-oxoadenosine, 8-Hydroxyadenosine, 8-Oxo-7,8-dihydroadenosine, 8-Oxyadenosine, 6-Amino-7,9-dihydro-9-beta-D-ribofuranosyl-8H-purin-8-one
IUPAC-namn (2R,3R,4S,5R)-2-(6-amino-8-hydroxypurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
LEDER Nc1ncnc2c1nc(O)n2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
Specifikationer
Kemiskt namn eller material 8-Oxoadenosine
Kvantitet 10 mg
InChI formel InChI=1S/C10H13N5O5/c11-7-4-8(13-2-12-7)15(10(19)14-4)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H,14,19)(H2,11,12,13)/t3-,5-,6-,9-/m1/s1
Formel vikt 283.0917