Läs mer
8-Quinolinemethanamine Dihydrochloride, TRC
Produktkod. 30424554
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30424554 1 g
1 options
Produktkod. 30424554 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ7007001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 8-Quinolinemethanamine Dihydrochloride
CAS 18004-63-2
Molekylformel C10H10N2.2HCl
Kvantitet 1 g
Synonym 8-(Aminomethyl)quinoline Dihydrochloride, [(Quinolin-8-yl)methyl]amine Dihydrochloride
InChI formel InChI=1S/C10H10N2.2ClH/c11-7-9-4-1-3-8-5-2-6-12-10(8)9;;/h1-6H,7,11H2;2*1H
LEDER NCc1cccc2cccnc12
Molekylvikt (g/mol) 231.12