Läs mer
(8S,10S)-10-[(3-Amino-2,3,6-trideoxy-Alpha-L-lyxo-hexopyranosyl)oxy]-8-(2-bromo-1,1-dimethoxyethyl)-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-5,12-naphthacenedione Hydrobromide(>85%)(Doxorubicin Impurity) Hydrobromide, TRC
Produktkod. 30328489
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30328489 10 mg
1 options
Produktkod. 30328489 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63040510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 13,13-Dimethoxy-14-Bromodaunorubicin hydrobromide
CAS 148218-14-8
Molekylformel C29 H34 Br N O11 . Br H
Kvantitet 10 mg
Synonym 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-8-(2-bromo-1,1-dimethoxyethyl)-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-, hydrobromide, (8S-cis)- (9CI), 13,13-Dimethoxy-14-bromodaunorubicin hydrobromide, (8S,10S)-10-[(3-Amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-8-(2-bromo-1,1-dimethoxyethyl)-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-5,12-naphthacenedione hydrobromide
InChI formel InChI=1S/C29H34BrNO11.BrH/c1-12-23(32)15(31)8-18(41-12)42-17-10-28(37,29(11-30,39-3)40-4)9-14-20(17)27(36)22-21(25(14)34)24(33)13-6-5-7-16(38-2)19(13)26(22)35;/h5-7,12,15,17-18,23,32,34,36-37H,8-11,31H2,1-4H3;1H/t12-,15-,17-,18-,23+,28-;/m0./s1
LEDER Br.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(CBr)(OC)OC
IUPAC-namn (7S,9S)-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-9-(2-bromo-1,1-dimethoxyethyl)-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrobromide
Molekylvikt (g/mol) 733.4
Formel vikt 731.0577