Läs mer
9,10-12,13-Diepoxyoctadecanoic Acid, TRC
Produktkod. 30373615
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30373615 25 mg
1 options
Produktkod. 30373615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44185525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 9,10-12,13-Diepoxyoctadecanoic Acid
CAS 3012-69-9
Molekylformel C18 H32 O4
Kvantitet 25 mg
Synonym 9,10:12,13-Diepoxyoctadecanoic Acid, 3-[(3-Pentyl-2-oxiranyl)methyl]-2-oxiraneoctanoic acid, 9,10:12,13-Diepoxyoctadecanoic acid, 9,12-Diepoxyoctadecanoic acid, Diepoxylinoleic acid, Linoleic acid dioxide, NSC 3581
InChI formel InChI=1S/C18H32O4/c1-2-3-7-10-14-16(21-14)13-17-15(22-17)11-8-5-4-6-9-12-18(19)20/h14-17H,2-13H2,1H3,(H,19,20)
LEDER CCCCCC1OC1CC2OC2CCCCCCCC(=O)O
IUPAC-namn 8-[3-[(3-pentyloxiran-2-yl)methyl]oxiran-2-yl]octanoic acid
Molekylvikt (g/mol) 312.444
Formel vikt 312.23