missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(phenylethynyl)anthracene, Thermo Scientific Chemicals

Catalog Number p-7058799
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10075-85-1
C30H18
378.474
MFCD00012050
ZHBOFZNNPZNWGB-UHFFFAOYSA-N
9,10-bis phenylethynyl anthracene, 9,10-bis 2-phenylethynyl anthracene, anthracene, 9,10-bis phenylethynyl, bpea, anthracene, 9,10-bis 2-phenylethynyl, bis phcc-anthracene, unii-fc8jdb70dq, 9,10-bis phenylvinyl anthracene, fc8jdb70dq, 9,10-bis-phenylethynyl anthracene
82338
CHEBI:51675
9,10-bis(2-phenylethynyl)anthracene
C1=CC=C(C=C1)C#CC2=C3C=CC=CC3=C(C4=CC=CC=C42)C#CC5=CC=CC=C5
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10075-85-1
C30H18
378.474
MFCD00012050
ZHBOFZNNPZNWGB-UHFFFAOYSA-N
9,10-bis phenylethynyl anthracene, 9,10-bis 2-phenylethynyl anthracene, anthracene, 9,10-bis phenylethynyl, bpea, anthracene, 9,10-bis 2-phenylethynyl, bis phcc-anthracene, unii-fc8jdb70dq, 9,10-bis phenylvinyl anthracene, fc8jdb70dq, 9,10-bis-phenylethynyl anthracene
82338
CHEBI:51675
9,10-bis(2-phenylethynyl)anthracene
C1=CC=C(C=C1)C#CC2=C3C=CC=CC3=C(C4=CC=CC=C42)C#CC5=CC=CC=C5

As a photochromic material exhibiting chemiluminescence

9,10-Bis(phenylethynyl)anthracene is used as a chemiluminescent fluorophore with high quantum efficiency. It is used in lightsticks as a fluorophor producing ghostly green light. It is also used as a dopant for organic semiconductors in OLEDs. It is used as a reagent for chemiluminescence research. 9,10-Bis(phenylethynyl)anthracene is known to exert fluorescence properties due to its polycyclic aromatic hydrocarbon system and it is commonly used as an scintillator additive. 9,10-Bis(phenylethynyl)anthracene was also studied extensively as a fluorescence emitters for peroxyoxalate chemiluminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 254°C to 255°C
Quantity 1 g
Beilstein 1891432
Sensitivity Light sensitive
Solubility Information It is insoluble in water but is soluble in most organic solvents such as carbon disulfide,alcohols,benzene,chloroform,and hydronaphthalenes.
Formula Weight 378.47
Physical Form Crystalline
Chemical Name or Material 9,10-Bis (phenylethynyl)anthracene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'