Läs mer
9,10-Dibromoanthracene-2-sulfonyl Chloride, TRC
Produktkod. 30376130
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376130 250 mg
30373188 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30376130 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD423500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 9,10-Dibromoanthracene-2-sulfonyl Chloride
CAS 210174-74-6
Molekylformel C14 H7 Br2 Cl O2 S
Kvantitet 250 mg
InChI formel InChI=1S/C14H7Br2ClO2S/c15-13-9-3-1-2-4-10(9)14(16)12-7-8(20(17,18)19)5-6-11(12)13/h1-7H
LEDER ClS(=O)(=O)c1ccc2c(Br)c3ccccc3c(Br)c2c1
IUPAC-namn 9,10-dibromoanthracene-2-sulfonyl chloride
Molekylvikt (g/mol) 434.53
Formel vikt 431.8222