Läs mer
9,10-Dichlorophenanthrene, TRC
Produktkod. 17941595
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17941595 5 mg
17932245 25 mg
17994755 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17941595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4353155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17219-94-2
Molekylformel C14 H8 Cl2
Molekylvikt (g/mol) 247.12
Synonym NSC 97575
IUPAC-namn 9,10-dichlorophenanthrene
LEDER Clc1c(Cl)c2ccccc2c3ccccc13
Specifikationer
Kemiskt namn eller material 9,10-Dichlorophenanthrene
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C14H8Cl2/c15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(13)16/h1-8H
Formel vikt 246.0003
Procent renhet >95%