Learn More
9,10-Diethoxyanthracene, 98%, Thermo Scientific Chemicals
206.70 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents.
9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 149°C to 151°C |
Quantity | 1 g |
Solubility Information | Soluble in toluene. |
Formula Weight | 266.33 |
Percent Purity | 98% |
Chemical Name or Material | 9,10-Diethoxyanthracene |