Läs mer
9,10-Dihydro-9-oxa-10-phosphaphenanthrene 10-Oxide, TRC
Produktkod. 30373671
Change view
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373671 2.5 g
30369339 500 mg
30379795 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30373671 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD6790332.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35948-25-5
Molekylformel C12H9O2P
Molekylvikt (g/mol) 216.17
Synonym 3,4:5,6-Dibenzo-2H-1,2-oxaphosphorin-2-oxide, 9,10-Dihydro-9-oxa-10-phosphaphenanthren 10-oxide, 9,10-Dihydro-9-oxa-10-phosphaphenanthrene 10-oxide, 9,10-Dihydro-9-oxa-10-phosphorylphenanthrene 10-oxide, Chemguard DOPO, DOP 11S25, DOPO, Epoclean, HCA, HCA (heat stabilizer), Hiretar 101, KFR-DOPO, Polydis 3710, Polyphlox 3742, Sanko HCA, Struktol Polydis 3710, Ukanol DOPO, Ukanol GKF, 9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide, Chemguard DOPO, DOPO, Epoclean, HCA, Hiretar 101, KFR-DOPO, Polydis 3710, Sanko HCA, Struktol Polydis 3710, Ukanol DOPO, 6H-Dibenz[c,e][1,2]oxaphosphorin 6-Oxide
IUPAC-namn benzo[c][2,1]benzoxaphosphinine 6-oxide
LEDER O=P1Oc2ccccc2c3ccccc13
Specifikationer
Kemiskt namn eller material 9,10-Dihydro-9-oxa-10-phosphaphenanthrene 10-Oxide
Kvantitet 2.5 g
InChI formel InChI=1S/C12H9O2P/c13-15-12-8-4-2-6-10(12)9-5-1-3-7-11(9)14-15/h1-8,15H
Formel vikt 216.034