missing translation for 'onlineSavingsMsg'
Learn More

9(10H)-Acridone, 99%, Thermo Scientific Chemicals

Catalog Number 11304629
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
578-95-0
C13H9NO
195.221
MFCD00005019
FZEYVTFCMJSGMP-UHFFFAOYSA-N
acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
2015
CHEBI:50756
10H-acridin-9-one
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2
This item is not returnable. View return policy
578-95-0
C13H9NO
195.221
MFCD00005019
FZEYVTFCMJSGMP-UHFFFAOYSA-N
acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
2015
CHEBI:50756
10H-acridin-9-one
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2

9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

Solubility
Insoluble in benzene, chloroform, ether, water and ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 360°C to 362°C
Quantity 1 g
Beilstein 7104
Solubility Information Insoluble in benzene,chloroform,ether,water and ethanol.
Formula Weight 195.22
Percent Purity 99%
Chemical Name or Material 9(10H)-Acridone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'