missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
9,11-Anhydro Fusidic Acid, TRC
Specifications
Specifications
| Kemiskt namn eller material | 9,11-Anhydrofusidic Acid |
| CAS | 74048-41-2 |
| Molekylformel | C31 H46 O5 |
| Kvantitet | 5 mg |
| Synonym | 29-Nordammara-9(11),17(20),24-trien-21-oic acid, 16-(acetyloxy)-3-hydroxy-, (3α,4α,8α,13α,14β,16β,17Z)- (9CI), ent-(17Z)-16α-(Acetyloxy)-3β-hydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-9(11),17(20),24-trien-21-oic acid, δ9(11)-Anhydrofusidic acid, (3alpha,4alpha,8alpha,13alpha,14beta,16beta,17Z)-16-(Acetyloxy)-3-hydroxy-29-nordammara-9(11),17(20),24-trien-21-oic Acid, Fusidic Acid EP Impurity L |
| InChI formel | InChI=1S/C31H46O5/c1-18(2)9-8-10-21(28(34)35)27-23-11-12-26-29(5)15-14-24(33)19(3)22(29)13-16-30(26,6)31(23,7)17-25(27)36-20(4)32/h9,12,19,22-25,33H,8,10-11,13-17H2,1-7H3,(H,34,35)/b27-21-/t19-,22-,23-,24+,25-,29-,30-,31-/m0/s1 |
| LEDER | C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)C2=CC[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C |
| IUPAC-namn | (2Z)-2-[(3R,4S,5S,8R,10S,13R,14S,16S)-16-acetyloxy-3-hydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,12,13,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid |
| Molekylvikt (g/mol) | 498.6939 |
| Formel vikt | 498.3345 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?