Läs mer
9,11-Anhydrobudesonide (Mixture of Diastereomers), TRC
Produktkod. 30135148
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135148 5 mg
30012289 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135148 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6894705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 313474-58-7
Molekylformel C25H32O5
Molekylvikt (g/mol) 412.52
Synonym Budesonide Imp. H (EP), 16alpha,17-[(1RS)-Butylidene-bis(oxy)]-21-hydroxypregna-1,4-diene-3,20-dione, (16alpha)-16,17-[Butylidenebis(oxy)]-21-hydroxy-pregna-1,4,9(11)-triene-3,20-dione, 9,11-Anhydrobudesonide, 16α,17-[(1RS)-Butylidenebis(oxy)]-21-hydroxypregna-1,4,9(11)-triene-3,20-dione
IUPAC-namn 16α,17-[(1RS)-Butylidenebis(oxy)]-21-hydroxypregna-1,4,9(11)-triene-3,20-dione
LEDER CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)C4=CC[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 16 alpha,17-[(1RS)-Butylidenebis(oxy)]-21-hydroxypregna-1,4,9(11)-triene-3,20-dione
Kvantitet 5 mg
InChI formel InChI=1S/C25H32O5/c1-4-5-22-29-21-13-19-17-7-6-15-12-16(27)8-10-23(15,2)18(17)9-11-24(19,3)25(21,30-22)20(28)14-26/h8-10,12,17,19,21-22,26H,4-7,11,13-14H2,1-3H3/t17-,19+,21-,22?,23+,24+,25-/m1/s1
Formel vikt 412.225