Läs mer
9(11)-Dehydrotestosterone Enanthate, TRC
Produktkod. 30364221
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30364221 100 mg
1 options
Produktkod. 30364221 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD230210100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 9(11)-Dehydrotestosterone Enanthate
CAS 119698-27-0
Molekylformel C26H38O3
Kvantitet 100 mg
Synonym 17beta-Hydroxyandrosta-4,9(11)-dien-3-one Heptanoate
InChI formel InChI=1S/C26H38O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h14,17,20-21,23H,4-13,15-16H2,1-3H3/t20-,21-,23-,25-,26-/m0/s1
LEDER CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)C3=CC[C@]12C
Molekylvikt (g/mol) 398.58
Formel vikt 398.2821