Läs mer
9-(2-Propenyl)adenine, TRC
Produktkod. 30419766
Change view
Kvantitet:
5 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419766 250 mg
30411381 5 mg
30411086 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30419766 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP768805250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4121-39-5
Molekylformel C8 H9 N5
Molekylvikt (g/mol) 175.19
Synonym 9-Allyladenine, N9-Allyladenine, NSC 77154, 9-(2-Propen-1-yl)-9H-purin-6-amine
IUPAC-namn 9-prop-2-enylpurin-6-amine
LEDER Nc1ncnc2c1ncn2CC=C
Specifikationer
Kemiskt namn eller material 9-(2-Propenyl)adenine
Kvantitet 250 mg
InChI formel InChI=1S/C8H9N5/c1-2-3-13-5-12-6-7(9)10-4-11-8(6)13/h2,4-5H,1,3H2,(H2,9,10,11)
Formel vikt 175.0858