Läs mer
9-[4-Acetoxy-3-(acetoxymethyl)butyl]-2-amino-6-chloropurine, Mikromol™
Produktkod. 30458637
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458637 4 x 25 mg
30051147 100 mg
30052467 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30458637 Leverantör Mikromol Leverantörsnummer MM0620.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
9-[4-Acetoxy-3-(acetoxymethyl)butyl]-2-amino-6-chloropurine, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 9-(4-Acetoxy-3-(acetoxymethyl)butyl)-2-amino-6-chloropurine
CAS 97845-60-8
Analyt- eller komponentnamn 9-(4-Acetoxy-3-(acetoxymethyl)butyl)-2-amino-6-chloropurine
Frakt skick Room Temperature
Molekylformel C14 H18 Cl N5 O4
Certifikat och efterlevnad ISO 17025
Synonym 1,3-Propanediol, 2-[2-(2-amino-6-chloro-9H-purin-9-yl)ethyl]-, 1,3-diacetate, 1,3-Propanediol, 2-[2-(2-amino-6-chloro-9H-purin-9-yl)ethyl]-, diacetate (ester) (9CI), 9-[4-Acetoxy-3-(acetoxymethyl)butyl]-2-amino-6-chloropurine, 6-Chloro Famciclovir, 6-Chlorofamciclovir
InChI formel InChI=1S/C14H18ClN5O4/c1-8(21)23-5-10(6-24-9(2)22)3-4-20-7-17-11-12(15)18-14(16)19-13(11)20/h7,10H,3-6H2,1-2H3,(H2,16,18,19)
Lösningstyp Neat
LEDER CC(=O)OCC(CCn1cnc2c(Cl)nc(N)nc12)COC(=O)C
IUPAC-namn [2-(acetyloxymethyl)-4-(2-amino-6-chloropurin-9-yl)butyl] acetate
Molekylvikt (g/mol) 355.78
Kvantitet 4 x 25 mg
Formel vikt 355.1047 g/mol