Läs mer
9-[4-Acetoxy-3-(acetoxymethyl)butyl]-2-amino-6-chloropurine, TRC
Produktkod. 30134107
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134107 10 g
30008600 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134107 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA16414010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97845-60-8
Molekylformel C14 H18 Cl N5 O4
Molekylvikt (g/mol) 355.78
Synonym 1,3-Propanediol, 2-[2-(2-amino-6-chloro-9H-purin-9-yl)ethyl]-, 1,3-diacetate, 1,3-Propanediol, 2-[2-(2-amino-6-chloro-9H-purin-9-yl)ethyl]-, diacetate (ester) (9CI), 9-[4-Acetoxy-3-(acetoxymethyl)butyl]-2-amino-6-chloropurine, 6-Chloro Famciclovir, 6-Chlorofamciclovir
IUPAC-namn [2-(acetyloxymethyl)-4-(2-amino-6-chloropurin-9-yl)butyl] acetate
LEDER CC(=O)OCC(CCn1cnc2c(Cl)nc(N)nc12)COC(=O)C
Specifikationer
Kemiskt namn eller material 9-(4-Acetoxy-3-(acetoxymethyl)butyl)-2-amino-6-chloropurine
Analyt- eller komponentnamn 9-(4-Acetoxy-3-(acetoxymethyl)butyl)-2-amino-6-chloropurine
Frakt skick Room Temperature
InChI formel InChI=1S/C14H18ClN5O4/c1-8(21)23-5-10(6-24-9(2)22)3-4-20-7-17-11-12(15)18-14(16)19-13(11)20/h7,10H,3-6H2,1-2H3,(H2,16,18,19)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 10 g
Formel vikt 355.1047