Learn More
9,9'-Spirobifluorene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.
Solubility
Soluble in common organic solvents. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.
Solubility
Soluble in common organic solvents. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 195°C to 200°C |
Color | White to Yellow |
Quantity | 250 mg |
Beilstein | 1914463 |
Solubility Information | Soluble in common organic solvents. Slightly soluble in water. |
Formula Weight | 316.4 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 9,9'-Spirobifluorene |