missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Acetylanthracene, 96%, Thermo Scientific Chemicals
395.26 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 15 - 30°C. Incompatible with oxidizing agents.
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 15 - 30°C. Incompatible with oxidizing agents.
Specifications
Melting Point | 72°C to 76°C |
Quantity | 5 g |
Beilstein | 1370056 |
Solubility Information | Insoluble in water. |
Formula Weight | 220.27 |
Percent Purity | 96% |
Chemical Name or Material | 9-Acetylanthracene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'