Learn More
9-Bromo-1-nonanol, 98%, Thermo Scientific Chemicals
Catalog Number p-7053744
Click to view available options
Quantity:
5 g
25 g
55362-80-6
C9H19BrO
223.15
MFCD00063348
USJDOLXCPFASNV-UHFFFAOYSA-N
9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq
108700
9-bromononan-1-ol
OCCCCCCCCCBr
55362-80-6
C9H19BrO
223.15
MFCD00063348
USJDOLXCPFASNV-UHFFFAOYSA-N
9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq
108700
9-bromononan-1-ol
OCCCCCCCCCBr
9-Bromo-1-nonanol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Bromo-1-nonanol is used as a pharmaceutical intermediate.
Solubility
Soluble in dichloromethane, ethyl acetate and hexane.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 33°C to 35°C
Boiling Point 125°C to 126°C (2 mmHg)
Flash Point 109°C (228°F)
Linear Formula Br(CH2)9OH
Quantity 5 g
Beilstein 1737525
Solubility Information Soluble in dichloromethane,ethyl acetate and hexane.
Formula Weight 223.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9-Bromo-1-nonanol