Läs mer
9-Bromo Hydrocortisone 21-Acetate, TRC
Produktkod. 30005332
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005332 10 mg
30009717 25 mg
30009897 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005332 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68447410MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50733-54-5
Molekylformel C23H31BrO6
Molekylvikt (g/mol) 483.39
Synonym (11beta)-21-(Acetyloxy)-9-bromo-11,17-dihydroxypregn-4-ene-3,20-dione, 9-Bromo-17-hydroxy Corticosterone 21-Acetate
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,17R)-9-bromo-11,17-dihydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(Br)[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material 9-Bromo Hydrocortisone 21-Acetate
Kvantitet 10 mg
InChI formel InChI=1S/C23H31BrO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h10,16-18,27,29H,4-9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1
Formel vikt 482.1304