missing translation for 'onlineSavingsMsg'
Läs mer

9-Bromo Hydrocortisone 21-Acetate, TRC

Produktkod. 30009897
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009897 50 mg
30005332 10 mg
30009717 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30009897 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68447450MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 50733-54-5
Molekylformel C23H31BrO6
Molekylvikt (g/mol) 483.39
Synonym (11beta)-21-(Acetyloxy)-9-bromo-11,17-dihydroxypregn-4-ene-3,20-dione, 9-Bromo-17-hydroxy Corticosterone 21-Acetate
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,17R)-9-bromo-11,17-dihydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(Br)[C@@H](O)C[C@]12C

Specifications

Kemiskt namn eller material 9-Bromo Hydrocortisone 21-Acetate
Kvantitet 50 mg
InChI formel InChI=1S/C23H31BrO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h10,16-18,27,29H,4-9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1
Formel vikt 482.1304
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.