missing translation for 'onlineSavingsMsg'
Learn More

9-Bromoanthracene, 96%, Thermo Scientific Chemicals

Catalog Number p-71730
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1564-64-3
C14H9Br
257.13
MFCD00001243
ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
74062
9-bromoanthracene
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br
This item is not returnable. View return policy
1564-64-3
C14H9Br
257.13
MFCD00001243
ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
74062
9-bromoanthracene
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 98°C to 100°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05, 665
Solubility Information Solubility in water: insoluble
Formula Weight 257.13
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 9-Bromoanthracene, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 9-Bromoanthracene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'