Läs mer
9-Chloro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Propanoate (Beclometasone 21-Propionate), Mikromol™
Produktkod. 18040590
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18040590 25 mg
18082690 100 mg
30457880 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18040590 Leverantör Mikromol Leverantörsnummer MM0199.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
9-Chloro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Propanoate (Beclometasone 21-Propionate)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclometasone 21-Propionate
CAS 69224-79-9
Analyt- eller komponentnamn Beclometasone 21-Propionate
Frakt skick Ice pack (-20°C)
Molekylformel C25 H33 Cl O6
Certifikat och efterlevnad ISO 9001
Synonym Beclometasone Dipropionate Imp. A (EP), 9-Chloro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Propanoate, Beclometasone 21-Propionate, (11beta,16beta)-9-Chloro-11,17-dihydroxy-16-methyl-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Beclomethasone Dipropionate Related Compound A, Beclometasone Dipropionate Impurity A
InChI formel InChI=1S/C25H33ClO6/c1-5-21(30)32-13-20(29)25(31)14(2)10-18-17-7-6-15-11-16(27)8-9-22(15,3)24(17,26)19(28)12-23(18,25)4/h8-9,11,14,17-19,28,31H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
LEDER CCC(=O)OCC(=O)[C@@]1(O)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molekylvikt (g/mol) 464.98
Kvantitet 25 mg
Formel vikt 464.1966 g/mol