Learn More
9-Chloroanthracene, 95%, BLD Pharm™
Product Code. 31430405
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31430405 25 g
31428057 250 mg
31419223 5 g
31429795 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31430405 Supplier BLD Pharm Supplier No. BD11171125g
 Restricted Item
Beräknad leverans: 25-11-2026
Specifications
CAS 716-53-0
Molecular Formula C14H9Cl
MDL Number MFCD00001245
Quantity 25 g
SMILES ClC1=C2C=CC=CC2=CC2=C1C=CC=C2
Molecular Weight (g/mol) 212.67
Percent Purity 95%
Chemical Name or Material 9-Chloroanthracene