Läs mer
9-Chlorophenanthrene (>90%), TRC
Produktkod. 30368302
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368302 100 mg
30367799 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368302 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC374975100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 947-72-8
Molekylformel C14 H9 Cl
Molekylvikt (g/mol) 212.67
Synonym Phenanthrene, 9-chloro-, 9-Chlorophenanthrene, 10-Chlorophenanthrene, NSC 8552
IUPAC-namn 9-chlorophenanthrene
LEDER Clc1cc2ccccc2c3ccccc13
Specifikationer
Kemiskt namn eller material 9-Chlorophenanthrene
Kvantitet 100 mg
InChI formel InChI=1S/C14H9Cl/c15-14-9-10-5-1-2-6-11(10)12-7-3-4-8-13(12)14/h1-9H
Formel vikt 212.0393