Läs mer
9-cis-Retinoic Acid Methyl Ester, TRC
Produktkod. 30425769
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30425769 25 mg
1 options
Produktkod. 30425769 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR24501025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 9-cis Retinoic Acid Methyl Ester
CAS 58526-50-4
Molekylformel C21 H30 O2
Kvantitet 25 mg
Synonym Retinoic acid','methyl ester','9-cis- (9CI), 9-cis-Methyl retinoate, 9-cis-Retinoic acid methyl ester, Methyl 9-cis-retinoate
InChI formel InChI=1S/C21H30O2/c1-16(9-7-10-17(2)15-20(22)23-6)12-13-19-18(3)11-8-14-21(19,4)5/h7,9-10,12-13,15H,8,11,14H2,1-6H3/b10-7+,13-12+,16-9-,17-15+
LEDER COC(=O)\C=C(/C)\C=C\C=C(\C)/C=C/C1=C(C)CCCC1(C)C
IUPAC-namn methyl (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoate
Molekylvikt (g/mol) 314.46
Formel vikt 314.2246