Läs mer
9-cis-Retinyl Linoleate, TRC
Produktkod. 30411719
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411719 10 mg
30422767 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30411719 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR27530010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79299-81-3
Molekylformel C38H60O2
Molekylvikt (g/mol) 548.88
Synonym 9-cis-Retinol Linoleate
IUPAC-namn [(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] (9Z,12Z)-octadeca-9,12-dienoate
LEDER CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(\C)/C=C/C1=C(C)CCCC1(C)C
Specifikationer
Kemiskt namn eller material 9-cis-Retinyl Linoleate
Kvantitet 10 mg
InChI formel InChI=1S/C38H60O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-27-37(39)40-32-30-34(3)25-22-24-33(2)28-29-36-35(4)26-23-31-38(36,5)6/h11-12,14-15,22,24-25,28-30H,7-10,13,16-21,23,26-27,31-32H2,1-6H3/b12-11-,15-14-,25-22+,29-28+,33-24-,34-30+
Formel vikt 548.4593