missing translation for 'onlineSavingsMsg'
Läs mer

9-cis-Retinyl Linoleate, TRC

Produktkod. 30422767
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422767 1 mg
30411719 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30422767 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR2753001MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79299-81-3
Molekylformel C38H60O2
Molekylvikt (g/mol) 548.88
Synonym 9-cis-Retinol Linoleate
IUPAC-namn [(2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] (9Z,12Z)-octadeca-9,12-dienoate
LEDER CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC\C=C(/C)\C=C\C=C(\C)/C=C/C1=C(C)CCCC1(C)C

Specifications

Kemiskt namn eller material 9-cis-Retinyl Linoleate
Kvantitet 1 mg
InChI formel InChI=1S/C38H60O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-27-37(39)40-32-30-34(3)25-22-24-33(2)28-29-36-35(4)26-23-31-38(36,5)6/h11-12,14-15,22,24-25,28-30H,7-10,13,16-21,23,26-27,31-32H2,1-6H3/b12-11-,15-14-,25-22+,29-28+,33-24-,34-30+
Formel vikt 548.4593
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.