Läs mer
9-Dehydrotestosterone, TRC
Produktkod. 30008801
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008801 10 mg
30011998 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008801 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23020510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2398-99-4
Molekylformel C19H26O2
Molekylvikt (g/mol) 286.41
Synonym 17beta-Hydroxyandrosta-4,9(11)-dien-3-one, 4,9(11)-Androstadiene-17beta-ol-3-one, 9(11)-Dehydrotestosterone, NSC 30932, U 3435
LEDER C[C@]12CC=C3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O
Specifikationer
Kemiskt namn eller material 9-Dehydrotestosterone
Kvantitet 10 mg
InChI formel InChI=1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h8,11,14-15,17,21H,3-7,9-10H2,1-2H3/t14-,15-,17-,18-,19-/m0/s1
Formel vikt 286.1933