Läs mer
9-Deschloro-9-bromo Beclomethasone Dipropionate, TRC
Produktkod. 30136233
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136233 10 mg -
30006030 2.5 mg 2.5mg
30011381 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136233 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28855510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52092-14-5
Molekylformel C28 H37 Br O7
Molekylvikt (g/mol) 565.49
Synonym (11β,16β)-9-Bromo-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Bromo-11β-hydroxy-16β-methyl-3,20-dioxopregna-1,4-diene-17,21-diyl Dipropanoate
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-bromo-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
LEDER CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Br)[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material 9-Deschloro-9-bromo Beclomethasone Dipropionate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C28H37BrO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1
Formel vikt 564.1723
Procent renhet >95%